Copper ≥99.999% (metals basis), wire, Ø 0.127 mm (0.005 in)

Supplier: Thermo Fisher Scientific

00100.G4 00100.G7
00100.G4EA 79.2 EUR
00100.G4 00100.G7
Copper ≥99.999% (metals basis), wire, Ø 0.127 mm (0.005 in)
Copper

Purity: 99.999% CAS No.: 7440-50-8 Physical Form: ≈0.11g/m unannealed Odour: Odourless MDL No.: MFCD00010965

Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR