Titanium ≥99.94% (metals basis), foil, Thickness: 0.025 mm (0.001 in)

Supplier: Thermo Fisher Scientific
Warning

00360.FH 00360.FL 00360.GT
00360.FHEA 106 EUR
00360.FH 00360.FL 00360.GT
Titanium ≥99.94% (metals basis), foil, Thickness: 0.025 mm (0.001 in)
Titanium

Purity: 99.94% CAS No.: 7440-32-6 Physical Form: ≈0.2g/25x75mm Odour: Odourless MDL No.: MFCD00011264

Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3262
Melting Pt: 1660
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR