Gold 99,9 Metals basis, wire, Ø 1.0 mm (0.04 in)

Supplier: Thermo Fisher Scientific

00727.BSEA 488 EUR
00727.BS 00727.BW
Gold 99,9 Metals basis, wire, Ø 1.0 mm (0.04 in)
Gold

Purity: 99.9% CAS No.: 7440-57-5 Physical Form: ≈15.15g/m Odour: Odourless MDL No.: MFCD00003436

Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR