Sodium hydroxide, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Caustic soda

0348.9025EA 634.11 EUR
0348.9025 0348.1000 0348.5000
Sodium hydroxide, BAKER ANALYZED® ACS, J.T.Baker®
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
Density: 2.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8 II
REACH: 01-2119457892-27
Merck Index: 14,08627

Order Now

Specification Test Results

Exceeds ACS Specifications
Assay min. 97.0 %
Calcium (Ca) max. 0.005 %
Chloride (Cl) max. 0.005 %
Potassium (K) max. 0.01 %
Magnesium (Mg) max. 0.002 %
Heavy Metals (as Ag) max. 0.001 %
Sodium Carbonate (Na2CO3) max. 1.0 %
Trace Impurities (in ppm)
Iron (Fe) max. 3
Mercury (Hg) max. 0.1
Nickel (Ni) max. 5
Nitrogen Compounds (as N) max. 3
Phosphate (PO4) max. 2
Sulfate (SO4) max. 5

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR