Sodium thiosulphate pentahydrate, high purity

Supplier: VWR Chemicals

0484-2.5KGEA 295 EUR
0484-2.5KG 0484-500G
Sodium thiosulphate pentahydrate, high purity
Sodium thiosulphate pentahydrate
Formula: Na₂S₂O₃.5H₂O
MW: 248.19 g/mol
Melting Pt: 48
Density: 1.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149186
CAS Number: 10102-17-7
EINECS: 231-867-5
Merck Index: 12,08844

Order Now

Specification Test Results

Insolubles <0.005%
pH (5%, Water) @25 °C 6.0 – 8.4
Purity >99%
Sulphate & Sulphite <0.1%
Sulphide <0.0001%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR