Ammonium acetate ≥97%, crystals, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor
Warning

Synonyms: Acetic acid ammonium salt

0596-07EA 0 EUR
0596-07 0596-R
Ammonium acetate ≥97%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Ammonium acetate
Formula: H₃CCOONH₄
MW: 77.08 g/mol
Melting Pt: 114 °C
Density: 1.17 g/cm³ (20 °C)
Flash Pt: 136 °C
Storage Temperature: Ambient
MDL Number: MFCD00013066
CAS Number: 631-61-8
EINECS: 211-162-9
Merck Index: 13,00495
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd160345483553318b94053de396335a9b18425f&vid=c6375b539008d6b43f50408d30b61261444faebc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use ≥97%
Assay (CH₃COONH₄) (by acid-base titration) ≥97 %
Insoluble Matter ≤0.005 %
Nitrate (NO₃) ≤0.001 %
pH of 5% Solution at 25°C 6.7 - 7.3
Residue after Ignition ≤0.010 %
Solution (20:100) - clear, colorless, complete Passes Test
Sulfate (SO₄) ≤0.001 %
Water (H₂O) ≤2.5 %
Chloride (Cl) ≤5 ppm
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm
Meets Reagent Specifications for testing USP/NF monographs

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR