Sodium nitrate, high purity
Supplier: VWR Chemicals
Warning
0598-500GEA 129 EUR
0598-500G
Sodium nitrate, high purity
Sodium nitrate
Formula: NaNO₃
MW: 84.99 g/mol
Boiling Pt: 380 °C (1013 hPa)
Melting Pt: 308 °C
Density: 2.26 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011119
CAS Number: 7631-99-4
EINECS: 231-554-3
UN: 1498
ADR: 5.1,III
REACH: 01-2119488221-41
Merck Index: 13,08720
Order Now
Specification Test Results
Insolubles ≤0.04%
Purity ≥99.0%