Sodium azide, high_purity

Supplier: VWR Chemicals
Danger

Synonyms: Sodium triazide

0639-250GEA 0 EUR
0639-250G
Sodium azide, high_purity
Sodium azide
Formula: NaN₃
MW: 65.01 g/mol
Melting Pt: 275
Density: 1.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003536
CAS Number: 26628-22-8
EINECS: 247-852-1
UN: 1687
ADR: 6.1 II
Merck Index: 13,08653

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR