Sodium iodide, Ultrapure

Supplier: VWR Chemicals
Warning

Synonyms: Hydriodic acid sodium salt, Sodium monoiodide, Ioduril

0853-250GEA 306 EUR
0853-250G 0853-500G
Sodium iodide, Ultrapure
Sodium iodide
Formula: NaI
MW: 149.89 g/mol
Boiling Pt: 1300
Melting Pt: 662
Density: 3.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003532
CAS Number: 7681-82-5
EINECS: 231-679-3
UN: 3077
ADR: 9 III
Merck Index: 13,08705

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR