VWR® Q Path® DC3, Decalcifier Rapid-Acting
Supplier: VWR Q-Path Chemicals
09128300.EA 96.9 EUR
09128300.
VWR® Q Path® DC3, Decalcifier Rapid-Acting
Stains Microscopy and staining reagents
Contains a mixture of hydrochloric acid and alcohols with some EDTA.
  • Decalcify larger tissues (already fixated)
  • Recommended volume is 20× the fragment volume
Ready to use for applications in histology and pathology laboratories.