Formic acid 94,5, high_purity

Supplier: VWR Chemicals
Danger

Synonyms: Aminic Acid, Methanoic acid

0961-100MLEA 81.8 EUR
0961-100ML 0961-500ML
Formic acid 94,5, high_purity
Formic acid
Formula: HCO₂H
MW: 46.03 g/mol
Boiling Pt: 100…101 °C (1013 hPa)
Melting Pt: 8.2…8.4 °C
Density: 1.22 g/cm³ (25 °C)
Flash Pt: 48
Storage Temperature: Ambient
MDL Number: MFCD00003297
CAS Number: 64-18-6
EINECS: 200-579-1
UN: 1779
ADR: 8 (3) II
REACH: 01-2119491174-37
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6cee80f8dc5a4afaf0be47215f1cf258eef7cfd&vid=a77e0bed84455e37172a184511237ad1d178fd11&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥94.5%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR