Potassium dihydrogen phosphate 99,999 Metals basis, Puratronic® Si_10ppm

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Monopotassiumphosphate, prim.-Potassium phosphate, Monopotassium phosphate, Potassium phosphate monobasic, Monobasic Potassium Phosphate, Potassium dihydrogen orthophosphate

10840.14 10840.22
10840.14EA 294 EUR
10840.14 10840.22
Potassium dihydrogen phosphate 99,999 Metals basis, Puratronic® Si_10ppm
Potassium dihydrogen phosphate

Purity: 99.999% CAS No.: 7778-77-0 Molecular Formula: H2KO4P Formula Weight: 136.09 Physical Form: Crystalline powder Density: 2.338 Appearance: White Melting Point: 253° Merck Reference: 14,7659 MDL No.: MFCD00011401

Formula: KH₂PO₄
MW: 136.09 g/mol
Boiling Pt: ∼100 °C (1013 hPa)
Melting Pt: 253
Density: 2.338
Storage Temperature: Ambient
MDL Number: MFCD00011401
CAS Number: 7778-77-0
EINECS: 231-913-4
REACH: 01-2119490224-41
Merck Index: 14,7659

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR