Copper 99,99 Metals basis, foil, Thickness: 1.0 mm (0.04 in)

Supplier: Thermo Fisher Scientific

10954.FI 10954.GB 10954.HC
10954.FIEA 71.2 EUR
10954.FI 10954.GB 10954.HC
Copper 99,99 Metals basis, foil, Thickness: 1.0 mm (0.04 in)
Copper

Purity: 99.99% CAS No.: 7440-50-8 Physical Form: ≈22.4g/50x50mm Odour: Odourless MDL No.: MFCD00010965

Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR