Disposable particle filtering respirators, FFP3, healthcare, Aura™ 1883+
Supplier: 3M
1883PLUS
111-1525EA 93.1 EUR
111-1525
Disposable particle filtering respirators, FFP3, healthcare, Aura™ 1883+
Respirators Disposable Respirators Particulate Respirators
Unique FFP3 respirator with two-way protection and added exhalation valve for comfort.
  • Three panel design for increased comfort with the added convenience of an individually wrapped foldable product
  • Includes shrouded valve fitted to the front of the respirator, which prevents heat and moisture build-up from exhaled breath
  • Meets requirements and testing for Bacterial Filtering Efficiency (BFE) and splash resistance
  • Red colour coded straps to denote FFP3 protection level
Certifikace: Highest Protection Level. Assigned Protection Factor = 20