Safety helmets, uvex pheos E-WR
Supplier: UVEX
9770.030 9770.130 9770.230 9770.530 9770.330 9770.430
111-2337EA 19.4 EUR
111-2337 111-2338 111-2339 111-2342 111-2340 111-2341
Safety helmets, uvex pheos E-WR
Head Protection Safety Helmets
The lightweight uvex pheos safety helmets feature a sporty design and offer maximum wearer comfort. The sturdy, rigid helmet shell also easily withstands pressure from the side, and fulfills all safety-related requirements for construction or industrial work.
  • Can quickly and easily be adapted to the user's individual needs - visor, safety spectacles, ear defenders or head torch
  • Suspension harness with wheel ratchet for variable width adjustment
  • Six-point textile band attachment ensures optimal fit and comfort
  • Climazone ventilation and long brim
  • With slot