Safety spectacles, Solus™ 2000 series
Supplier: 3M
S2001AF-BLU S2002SGAF-BGR S2003SGAF-BGR S2005SGAF-BGR S2001ASP-BLU S2007SGAF-BGR S2024AS-RED S2001SGAF-BGR S2001SGAF-RED S2001SGAFBGRF
111-2499EA 17.5 EUR
111-2499 MMMAS2002SGAF-BGR MMMAS2003SGAF-BGR MMMAS2005SGAF-BGR 111-2502 MMMAS2007SGAF-BGR MMMAS2024AS-RED MMMAS2001SGAF-BGR MMMAS2001SGAF-RED MMMAS2001SGAFBGRF
Safety spectacles, Solus™ 2000 series
Safety Glasses
Comfortable, stylish eyewear with a vast array of colour and lens tint options, the Solus™ 2000 series provides eye protection with a stylish flare.
  • Adjustable ratchet temples
  • Scotchgard™ anti-fog coating resists fogging longer than traditional anti-fog coatings
  • Polycarbonate lenses absorb 99,9% of UVA and UVB rays
The Solus™ 2000 series features adjustable ratchet temples that allow the wearer to customise the fit. Choose from several lens coating options, including Scotchgard™ anti-fog, multiple lens colours, and features like a removable foamMore Product Information