Powered and supplied air breathing tubes, Versaflo™
Supplier: 3M
BT-64
111-2553EA 384 EUR
111-2553
Powered and supplied air breathing tubes, Versaflo™
Respirators Respiratory Systems
The Versaflo™ Breathing Tube BT-64 is a heavy-duty breathing tube. It connects approved tight fitting facepieces to powered air delivery units to ensure that clean air reaches the facepieces with unrestricted airflow.
  • Made from heavy-duty material
  • Suitable for demanding applications
  • Protects against certain gases, vapours, and particulates when used in an approved configuration
  • Airflow helps provide comfort whilst working and helps reduce visor fogging
  • Supports integration of compatible hearing and head protection
  • Fully submersible for cleaning (with 3M™