Cleanroom gloves, nitrile, sterile, Kimtech™ G3 Sterling™
Supplier: KIMBERLY CLARK
11821 11822 11823 11824 11825 11826 11827 11828
112-1858EA 603 EUR
112-1858 112-1859 112-1860 112-1861 112-1862 112-1863 112-1864 112-1865
Cleanroom gloves, nitrile, sterile, Kimtech™ G3 Sterling™
Gloves
Hand-specific, sterile gloves with beaded cuff for cleanroom applications. When double donned, these gloves deliver the equivalent feel and dexterity of a single latex glove without the risk of type I latex allergic reactions.
  • Sterile, powder-free, textured finish
  • Environmentally responsible thanks to improved packaging that saves waste
  • Static dissipative in use
  • Comfortable fit, suitable for double donning
  • Particles per cm² ≥0,5 micron: max. 1200
Suitable for use in Class 1, ISO 3 or M1.5 or higher cleanrooms.
Length: 300 mm
Thickness: 0,08 mm (More Product Information