Latex gloves, Microflex® 63-864
Supplier: Ansell
63-864/8.5-9 63-864/9.5-10 63-864/6.5-7 63-864/5.5-6 63-864/7.5-8
112-4453EA 36.7 EUR
112-4453 112-4454 112-4451 112-4450 112-4452
Latex gloves, Microflex® 63-864
Gloves
These disposable gloves are thicker than average Ansell latex gloves and provide chemical and liquid protection.
  • Non sterile, powder-free
  • Unique design that prevents rips and tears
  • Enhanced, textured fingertips provide a secure, dependable grip of dry and wet parts
  • Palm thickness meets and exceeds quality standards
  • Pinhole rates are lower than the standard minimum requirements for average latex gloves
Ideal for use in automotive applications, for assembly and inspection, food processing, general maintenance, painting and spraying, cleaningMore Product Information