VWR® Advanced, Cut Resistant Gloves
Supplier: VWR Collection
112-5685EA 50.7 EUR
112-5685 112-5686 112-5687 112-5688 112-5689 112-5690
VWR® Advanced, Cut Resistant Gloves
Gloves
The VWR® Advanced glove with polyurethane coating provides enhanced cut protection for a range of applications that would include a high risk of cuts.
  • ANSI A3 Cut Protection (Level C)
  • Nylon/spandex/polyester blend composition
  • Polyurethane (PU) coating on palm and fingertips
  • Grey knitted gloves with black coating
Ideal for handling items or tools with a high risk of cuts, including handling of items with sharp or jagged edges, assembly, maintenance, and janitorial applications.
Certifikace: Meets requirements for ANSI A3 Cut Resistance. EN 388:2016 (More Product Information