Cleanroom gloves, latex, sterile, Kimtech™ G3
Supplier: KIMBERLY CLARK
56843 56844 56845 56846 56847 56848 56849 56842
112-6413EA 390 EUR
112-6413 112-6412 112-6411 112-6410 112-6409 112-6408 112-6407 112-6414
Cleanroom gloves, latex, sterile, Kimtech™ G3
Gloves
These sterile latex gloves are designed to ensure high levels of contamination control in critical and controlled cleanroom environments.
  • High levels of contamination control in critical and controlled cleanroom environments
  • Sterilised latex ensures compliant and seamless protection when and where it counts
  • High-quality finish, with enhanced texturing on the palm and fingertips, provides improved grip, tactile sensitivity and comfort in both wet and dry conditions
  • Hand-specific gloves also feature a beaded cuff that provides added strength and makes
More Product Information
Order Now
SPECIFICATIONS
Ambidextrous / hand-specific Hand-specific
Environmentally Preferable
Disposable / reusable Disposable
Latex / latex-free Latex
Powdered / powder-free Powder-free
Cleanroom class 100/ISO 5; EU GMP (A)
Finish Textured palm and fingertips
Cuff style Beaded
Cuff thickness 0,14 mil
Finger thickness 0,22 mil
Palm thickness 0,20 mil
Tensile strength 28 MPa
EN norm EN ISO 374-1:2016 Type C (K), EN 374-4:2003, EN ISO 374-5:2016 VIRUS