Beard cover, Hopen®
Supplier: Medicom
70.801
113-8337EA 16 EUR
113-8337
Beard cover, Hopen®
Disposable Garments
This disposable beard cover made of white PP covers the lower part of the face. It is HACCP approved and therefore ideal for use in food applications, such as catering and food processing. Also suitable for chemical, pharmaceutical and maintenance applications.
  • Sewn, elasticated
  • Latex-free
Weight: 1,5 g
Size: 45 cm (long, flat)