Iron 99,994 Metals basis, foil, Puratronic®, Thickness: 0.5 mm (0.02 in)

Supplier: Thermo Fisher Scientific

11379.FF 11379.FI 11379.GH
11379.FFEA 145 EUR
11379.FF 11379.FI 11379.GH
Iron 99,994 Metals basis, foil, Puratronic®, Thickness: 0.5 mm (0.02 in)
Iron

Purity: 99.994% CAS No.: 7439-89-6 Physical Form: ≈2.5g/25x25mm MDL No.: MFCD00010999

Formula: Fe
MW: 55.85 g/mol
Boiling Pt: 3000
Melting Pt: 1535
Density: 7.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010999
CAS Number: 7439-89-6
EINECS: 231-096-4
Merck Index: 13,05109

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR