Gold ≥99.9985% (metals basis), foil, Premion®, Thickness: 1.0 mm (0.04 in)

Supplier: Thermo Fisher Scientific

11388.FF 11388.FI
11388.FFEA 3330 EUR
11388.FF 11388.FI
Gold ≥99.9985% (metals basis), foil, Premion®, Thickness: 1.0 mm (0.04 in)
Gold

Purity: 99.9985% CAS No.: 7440-57-5 Physical Form: ≈12g/25x25mm Odour: Odourless MDL No.: MFCD00003436

Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR