VWR®, Occupational shoes, clogs, autoclavable
Supplier: VWR Collection
114-5004EA 0 EUR
114-5004 114-5002
VWR®, Occupational shoes, clogs, autoclavable
Shoes and Boots
Anti-static clogs made from thermoplastic material, with a wide fitting, comfortable shape and a slip resistant sole. Machine washable at 90 °C; autoclavable at 135 °C.
  • Resistant to high temperatures
  • Energy absorption heel
  • Side perforations for ventilation
  • Antibacterial, anti-vibration injected insole
  • Attached pivoting heel strap
Colour: White, black, green, blue, pink or red
Colour: White, black, green, blue.
Certifikace: EN ISO 20347; OB, A, E, SRB.