VWR®, Cleanroom clogs
Supplier: VWR Collection
114-8980EA 170 EUR
114-8980 114-8981 114-8982 114-8983 114-8984 114-8985
VWR®, Cleanroom clogs
Shoes and Boots
VWR® cleanroom clogs provide comfort and durability.
  • Anti-static
  • Autoclavable
  • Smooth surface for day long comfort
  • Special conical design prevents spilled liquids from entering clog
  • Heel strap
  • Non slip sole
  • Anatomic heel basin
They have been designed with foot comfort in mind. Smooth surface and aeration holes on both sides provide day extended comfort. Special conical design prevents spilled liquids from entering inside the clog.
These shoes are suitable for autoclave sterilisation. They can also be cleaned in 100 °C with domestic detergents,More Product Information