Silver ≥99.9% (metals basis), foil, annealed, Thickness: 0.127 mm (0.005 in)

Supplier: Thermo Fisher Scientific

11440.FIEA 61.5 EUR
11440.FI 11440.GH 11440.GW
Silver ≥99.9% (metals basis), foil, annealed, Thickness: 0.127 mm (0.005 in)
Silver

Purity: 99.9% CAS No.: 7440-22-4 Physical Form: ≈3.3g/50x50mm, Temper: soft Odour: Characteristic MDL No.: MFCD00003397

Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212
Melting Pt: 962
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR