Nickel 99,5 Metals basis, rods, Ø 6.35 mm (0.25 in)

Supplier: Thermo Fisher Scientific
Danger

11454.BV 11454.VB
11454.BVEA 107 EUR
11454.BV 11454.VB
Nickel 99,5 Metals basis, rods, Ø 6.35 mm (0.25 in)
Nickel

Purity: 99.5% CAS No.: 7440-02-0 Physical Form: ≈2.82g/cm Odour: Odourless MDL No.: MFCD00011137

Formula: Ni
MW: 58.69 g/mol
Boiling Pt: 3007
Melting Pt: 1453
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1 III
Merck Index: 12,06582

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR