Cleanroom wipes, sterile, SterileWipe™ AS 10
Supplier: Texwipe
TX3215S
115-1283EA 296 EUR
115-1283
Cleanroom wipes, sterile, SterileWipe™ AS 10
Wipes Cleanroom Wipes Sterile Cleanroom Wipes
SterileWipe™ AS 10 is a 2 ply, double knit construction, 100% continuous filament polyester wipe that offers high sorbency in a sealed-border wipe to prevent particle and fibre release.
  • Extremely low levels of releasable particles and fibres
  • Large wipe size and liquid capacity simplify the application of large volumes of cleaning and disinfecting solutions
  • Withstands exposure to common cleaning and disinfecting solutions including bleaches, phenols and quaternary ammonium compounds
  • Sterilised by gamma radiation to a 10⁻⁶ SAL (Sterility Assurance Level) in
More Product Information
Order Now
SPECIFICATIONS
Model SterileWipe™ AS 10
Environmentally Preferable
Basis weight 195 g/m²
Absorbency 595 ml/m²
Absorbency rate 0,3 s
Extractables - using deionised water 0,05 g/m²
Extractables - using isopropyl alcohol 0,09 g/m²
Fibres (>100 µm) 230/m²
Particle generation (0,5 - 5 µm) - ASTM E2090 test 6 × 10⁶/m²
Particle generation (5 - 100 µm) - ASTM E2090 Test 315000/m²