Cleanroom wipes, pre-saturated, sterile AlphaSat® 10
Supplier: Texwipe
TX3285
115-1891EA 1480 EUR
115-1891
Cleanroom wipes, pre-saturated, sterile AlphaSat® 10
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
This 100% continuous filament polyester wipe with thermally sealed borders is engineered to have the lowest level of releasable particles and fibres for critical and sterile environments. The wipe is pre-wetted with 70% USP IPA.
  • Optimises IPA use for effective particulate removal and eliminates overspray of alcohol
  • Superior strength for rigorous wiping without snagging on equipment
  • Wipe glides smoothly over surfaces with low abrasion
  • Sized appropriately for limited isolator space
Ideal for cleaning cleanroom environments including the interiors ofMore Product Information