Wipes for delicate tasks, Kimtech™ Science
Supplier: KIMBERLY CLARK
7551 7552 7552_CH 7551_CH 7557 7558
115-2029EA 719 EUR
115-2029 115-2221 115-2093 KIMB7551_CH 115-2074 115-2075
Wipes for delicate tasks, Kimtech™ Science
Wipes Disposable Wipes and Towels Wipes for Delicate Tasks
These high quality wipes, made from 100% pure fibre, are suitable for a variety of precision wiping tasks such as the cleaning of delicate glassware, equipment and surfaces. They are also suitable for cleaning liquids and dust, and are equipped with anti-static dispensing to reduce lint and anti-static discharge.
  • Light-duty wipes
  • Low lint absorbency for delicate applications and surfaces
  • Chemically inert and anti-static
  • Supplied in a hygienic, user friendly pop-up box for clean and fast dispensing, even while on the move
Ideal for applications inMore Product Information