Protective hand and body cream, Stokoderm® Protect PURE
Supplier: Deb-STOKO
UPW100ML UPW1L
115-2386EA 49.44 EUR
115-2386 115-2387
Protective hand and body cream, Stokoderm® Protect PURE
Skincare Skin Creams Hand Creams
Universal pre-work cream to help protect the skin against water and non water-based workplace contaminants, such as oils and solvents.
  • With glycerine and conditioner with skin protecting material
  • Silicone-free, perfume-free
Food industry compatible. Suitable for use on hands, face, and body. Also suitable for use with gloves.
Upozornení: *The product(s) on this page are outside the scope of the EU Directive 2019/904