Scourers, PurActive
Supplier: FHP VILEDA SCS
123111_U 123112_U 123113_U 123114_U 123111 123112 123113 123114
129-0856EA 14.3 EUR
129-0856 129-0857 129-0858 129-0859 115-2575 115-2576 115-2577 115-2578
Scourers, PurActive
Wipes Sponges
Scrubber sponges made of polyurethane foam.The special coating easily lifts stubborn grime whilst being gentle on all hard surfaces.
  • Foam backing is soft to the hands and provides a good grip
  • Removes dirt effectively without scratching
  • Colour-coding system enables cleaning process according to hygiene standards and HACCP principles
  • Washable up to 60 °C
The scourer cleans three times more effectively than traditional non-scratching scourers and is more gentle on surfaces than a traditional abrasive scourer.