Mop wipe systems, VertiKlean® MAX®
Supplier: CONTEC
LWMM0300 LWQT0100 PSQT0100
115-1861EA 733 EUR
115-1861 115-3201 115-3202
Mop wipe systems, VertiKlean® MAX®
Mops
The VertiKlean® MAX Mop Wipe Systems offer a quick and easy solution to mopping floors, walls, and other flat surfaces in any controlled environment. These versatile systems work with a variety of mop wipe covers, enabling the user to customize the cleaning task.
  • Stainless steel frame is chemical resistant
  • Easy to attach and remove mop covers
  • Variety of fabric options available
  • Removes contaminants easily from the environment
  • Slim profile allows easy access to confined spaces for efficient cleaning and disinfection
  • Thin, lightweight design makes it easy to