Sampling swabs
Supplier: DELTALAB
300205 300260 300232 300230
115-8231EA 40.9 EUR
115-8231 115-8232 115-8250 DELT300230
Sampling swabs
Swabs Swabs for Sampling
For safe sampling. Item 115-8250 is longer than conventional swabs and was designed to reach hard-to-reach areas. For example, suitable for use in gynaecology when used with a speculum to remove endocervical cells. Also ideal for veterinary medicine.
  • All swabs have round, tight, tear-resistant heads to ensure perfect sampling without residues
Tip Ø: 5 mm
Upozornení: *The product(s) on this page are outside the scope of the EU Directive 2019/904