Washdown resistant labels, ToughWash™
Supplier: Brady
711224 711225 711226 711227 711228 711229 711230 711232 711241 711242 711246 711248 711250 711259 711260
116-0404EA 206.85 EUR
116-0404 116-0405 116-0406 116-0407 116-0402 116-0403 116-0408 116-0410 116-0411 116-0412 116-0392 116-0394 116-0395 116-0397 116-0398
Washdown resistant labels, ToughWash™
Labels
The Toughwash™ washdown resistant labels are a durable, print on demand pressure sensitve adhesive product.
  • Protection from harsh washdown environments
  • Tested at washdown temperatures up to 80 °C and pressures up to 1500 psi
Ideal for the food and beverage processing industry.