Potassium iodide 99,0 ACS

Supplier: Thermo Fisher Scientific

Synonyms: Dipotassium diiodide, Kali iodide

11601.22 11601.36 11601.A3
11601.22EA 59.4 EUR
11601.22 11601.36 11601.A3
Potassium iodide 99,0 ACS
Potassium iodide

Purity: 99.0% min CAS No.: 7681-11-0 Molecular Formula: KI Formula Weight: 166.01 Physical Form: Granular Density: 3.12 Refractive Index: 1.677 Sensitivity: Hygroscopic Odour: Odourless Melting Point: 681° Boiling Point: 1330° Solubility: Soluble in water, alcohol, acetone, and glycerol Merck Reference: 14,7643 MDL No.: MFCD00011405


In photographic emulsions, in analytical chemistry, as a feed additive, in spectroscopy, in infrared transmission, and in scintillation

Formula: Kl
MW: 166 g/mol
Boiling Pt: 1330 °C
Melting Pt: 681 °C
Density: 3.12
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0
EINECS: 213-659-4
Merck Index: 14,7643

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR