Tech pens, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F133840001
119-0442EA 59.1 EUR
119-0442
Tech pens, SP Bel-Art
Pens Ink Pens
Heavy-duty soft tip disposable pen cornes in several colours and write on glass, metal, ceramics, plastics, paper etc. The oil-based marking paint resists washing, weathering, mild acids and alkalis. mark is app. 1 mm wide.
  • Writes on most surfaces
  • Operates within the temperature range of −46 to +65 °C
  • Autoclavable at 121 °C