Zinc ≥99.994% (metals basis), wire, Puratronic®, Ø 0.5 mm (0.02 in)

Supplier: Thermo Fisher Scientific

12054.G5 12054.G1
12054.G5EA 190 EUR
12054.G5 12054.G1
Zinc ≥99.994% (metals basis), wire, Puratronic®, Ø 0.5 mm (0.02 in)
Zinc

Purity: 99.994% CAS No.: 7440-66-6 Physical Form: ≈1.40g/m Odour: Odourless MDL No.: MFCD00011291

Formula: Zn
MW: 65.39 g/mol
Boiling Pt: 908
Melting Pt: 420
Density: 7.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR