Disposable spill kit
Supplier: Brady
813868
121-0136EA 93.06 EUR
121-0136
Disposable spill kit
Spill Control Materials
Designed for small spills of chemicals and hazardous liquids up to 1,7 L. The highly visible dispenser box can easily be be wall or shelf mounted.
  • Disposable brush to sweep up any broken glass or debris
  • Protective gloves have roughened finger tips, increased tear resistance and good chemical resistance
  • Autoclavable disposal bag
Delivery information: Kit contains two absorbent pads (410×510 mm), one pair of disposable chemical protective gloves (powder free), disposal bag, one disposable brush, and multilingual instruction guide.