PIG® HazMat chemical absorbent socks
Supplier: NEW PIG
124CR PIG301
121-0501EA 140.5 EUR
121-0501 121-0538
PIG® HazMat chemical absorbent socks
Spill Control Materials
Stop the spread of acids and caustics with these specially treated socks. Ideal for highly concentrated corrosives.
  • Absorbs the widest range of acids, caustics and unknown liquids, even those with high concentrations like 98% sulfuric acid and 30% sodium hydroxide
  • Chemical-resistant sock won't degrade or cause a dangerous reaction upon contact with corrosive spills
  • Excellent for quickly diking and stopping the spread of chemical spills; for diking larger chemical spills, consider our larger diameter PIG HazMat Dikes
  • PP skin resists chemicals and tearing;