Palladium(II) chloride solution_SC (9,0 - 11,0% Pd) w/w

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Palladium dichloride

12623.NB 12623.NH
12623.NBEA 827 EUR
12623.NB 12623.NH
Palladium(II) chloride solution_SC (9,0 - 11,0% Pd) w/w
Palladium (II) chloride

CAS No.: 7647-10-1 Molecular Formula: PdCl2 Formula Weight: 177.31 Physical Form: Liquid Appearance: Dark brown Odour: Acidic UN No.: UN3264 MDL No.: MFCD00003558

Formula: PdCl₂
MW: 177.33 g/mol
Melting Pt: 678
Storage Temperature: Ambient
MDL Number: MFCD00003558
CAS Number: 7647-10-1
EINECS: 231-596-2
UN: 3264
ADR: 8 II
Merck Index: 13,07058

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR