Intervoid® tamper evident mailing bags
Supplier: COVERIS
COV-512
129-1030EA 868 EUR
129-1030
Intervoid® tamper evident mailing bags
Bags Sample Bags
Intervoid® tamper evident mailing bags are designed to ensure the secure dispatch of samples or confidential data by mail courier. These respond to the critical needs of having a real guaranty of traceability, confidentiality and security when transporting samples.
  • Opaque polyethylene film protects the shipment from light and provides confidentiality
  • Traceability: one barcode with sequential numbering and detachable receipt
  • High-security: closure, unique marking system (pat. Pend.), micro printing