BinLiner™ Stands™
Supplier: MICRONOVA
BLSSH-40 BLSSH-1624 BLSSH-1515 BLSSH-168
129-1585EA 560 EUR
129-1585 129-1586 MICRBLSSH-1515 MICRBLSSH-168
BinLiner™ Stands™
Racks Bag Racks
The stainless steel Micronova BinLiner™ Stands™ are designed to work flawlessly with the BinLiners™ and similar sized bags. Securely hold a range of bin liner bags and feature two styles of open rod frames that allow ergonomic side removal of liners for simplified use.
  • Autoclavable and resistant to common cleaning solutions and disinfectants
The unique rectangular stainless steel wire stand includes plastic corners and feet to hold waste and garment disposal type bags/liners. The plastic feet (polyketone moulds) move easily on cleanroom floors withoutMore Product Information