Slim Jim® Step-On Containers
Supplier: Rubbermaid Commercial Products
1883557 1883566 1883573 1883555 1883579 1883606 1883575 1883552 1883615 1883613 1883611 1883609 1883459 1883458 1883460 1883553 1883456 1883559 1883577 1883551 1883602 1883604 1883600 1883568 1883563 1883564 1883455 1883554 1883561 1883570
129-2026EA 282 EUR
129-2026 129-2067 129-2069 129-2025 129-2073 129-2078 129-2071 129-1992 129-2074 129-2083 129-2081 129-2080 129-2183 129-2184 129-2056 129-2021 129-2186 129-2107 129-2072 129-1989 129-2076 129-2077 129-2075 129-2068 129-2064 129-2065 129-2187 129-3984 129-3983 129-3979
Slim Jim® Step-On Containers
Bins Waste Bins
Safely collect waste wih touch-free closed lid containers.
  • Premium quality construction ensures product durability in demanding environments
  • Slim profile and small footprint maximise space to fit in tight spaces
  • Internal hinge design prevents wall damage
  • Quiet and controlled lid-closure minimises noise to enhance guest and patient satisfaction
  • Smooth surfaces and contoured edges are easy to clean
  • Available in multiple sizes, formats, and colours
The Rubbermaid Commercial Slim Jim® Step-On Container has a small footprint and offers increased pedalMore Product Information