Legacy Step-On Containers
Supplier: Rubbermaid Commercial Products
FG614300RED FG614400WHT FG614400RED FG614500RED FG614600WHT FG614300WHT FG614600RED FG614300BEIG FG614400BEIG FG614600BEIG FG614500WHT
129-3512EA 216 EUR
129-3512 129-3521 129-3709 129-3532 129-3716 129-0395 129-1961 129-3967 129-3962 129-3975 RUBBFG614500WHT
Legacy Step-On Containers
Bins Waste Bins
These Rubbermaid Commercial step-on containers provide sanitary waste management.
  • Slow close, integrated lid damper keeps odours and bacteria from escaping
  • Puncture-resistant, all-plastic construction
  • Ideal for medical waste containment
  • Quiet, heavy-duty pedal for hands-free, sanitary use
  • Compliant with industry standards, such as FM, OSHA, and OBRA
  • Supplied with a five-year warranty
The Legacy step-on container enables hands-free sanitary waste management. These trash cans are ideal for areas where medical waste originates, including patient rooms,More Product Information