Chemistry vacuum pumps and systems with valve control, dual application
Supplier: VACUUBRAND
20737351 20733150 20733151 20733152 20733250 20733251 20733252 20733350 20733351 20733352 20737150 20737151 20737152 20737250 20737251 20737252 20737350 20737352
131-0044EA 0 EUR
131-0044 131-0020 131-0021 131-0022 131-0023 131-0024 131-0025 131-0026 131-0027 131-0028 131-0037 131-0038 131-0039 131-0040 131-0041 131-0042 131-0043 131-0045
Chemistry vacuum pumps and systems with valve control, dual application
Pumps Vacuum Diaphragm Pumps
These chemistry pumping units use chemical resistant solenoid valves to maintain process control. Once the first boiling point is detected by the controller, it maintains the pressure at this level. Pumping units can be configured to run one or two independent processes at the same time.
  • VACUU·SELECT vacuum controller with graphical user interface simplifies work in the laboratory
  • Predefined applications save time and give reproducible results
  • Easy-to-use application editor allows own process sequences
  • Excellent environmental friendliness due to efficient