Safety storage cabinets, Type 90, Q-CLASSIC-90
Supplier: asecos
30001040 30001041 30001046 30001047 30001048 30001049 30001050 30041040 30041041 30041046 30041047 30041048 30041049 30041050 30072040 30072041 30072046 30072047 30072048 30072049 30072050 30073040 30073041 30073046 30073047 30073048 30073049 30073050 37586040 3000104033640 37587040
135-2181EA 3590 EUR
135-2181 135-1742 135-2182 135-2183 135-2184 135-2185 135-2186 135-1749 135-2270 135-2366 135-2367 135-2368 135-2369 135-2370 135-2242 135-1753 135-2388 135-2389 135-2390 135-2391 135-2392 135-2393 135-2394 135-2395 135-2396 135-2397 135-2398 135-2399 MSGA37586040 MSGA3000104033640 MSGA37587040
Safety storage cabinets, Type 90, Q-CLASSIC-90
Furniture Storage Cabinets
Fireproof safety cabinets, classified Type 90 (fire resistant up to 90 minutes), safe, durable, robust and convenient with transport base and bi-coloured cabinet design.
  • Robust construction and longevity - triple-hinged door, safety elements assembled outside the storage compartment for increased protection against corrosion, scratch and impact resistant surface, easy to clean
  • Easy handling with comfort - doors stop in any position, self-closing in the event of fire
  • No unauthorised use - doors lockable with cylinder locking (integration in a locking system