Tin(IV) iodide, dry_ultra 99,998 Metals basis

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Tin tetraiodide

ampuled under argon

13617.06 13617.14
13617.06EA 254 EUR
13617.06 13617.14
Tin(IV) iodide, dry_ultra 99,998 Metals basis
Tin (IV) iodide

Purity: 99.998% CAS No.: 7790-47-8 Molecular Formula: SnI4 Formula Weight: 626.31 Physical Form: Powder, Ampouled under argon Density: 4.46 Refractive Index: 2.106 Sensitivity: Moisture Sensitive Melting Point: ca 143° Boiling Point: 340° Merck Reference: 14,8777 UN No.: 3260 MDL No.: MFCD00036272

Formula: I₄Sn
MW: 626.33 g/mol
Boiling Pt: 340 °C
Melting Pt: ∼ 143 °C
Density: 4.46
Storage Temperature: Ambient
MDL Number: MFCD00036272
CAS Number: 7790-47-8
EINECS: 232-208-4
UN: 3260
ADR: 8 III
Merck Index: 14,8777

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR