Selenium standard solution, 1000 mg/L Se in 5% nitric acid (from Se), standard for ICP, Specpure®

Supplier: Thermo Fisher Scientific

13845.AD 13845.AE 13845.AP
13845.ADEA 57.7 EUR
13845.AD 13845.AE 13845.AP
Selenium standard solution, 1000 mg/L Se in 5% nitric acid (from Se), standard for ICP, Specpure®
Standards Cationic Standards Selenium Standards

Molecular Formula: Se in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134090


Certifikace: Traceable to NIST

Order Now

SPECIFICATIONS

Environmentally Preferable
MDL No. MFCD00134090

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR